missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Microglass Fiber Filter Discs

Made with 100% borosilicate micro-glass fibers. Fisherbrand™ Microglass Fiber Filter Discs are non-hygroscopic and biologically inert. They show chemical resistance to most solvents and reagents (except hydrofluoric acid).

Brand:  Fisherbrand™

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11778553

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

  • Contains no binder
  • Suitable for use at and up to 500°C
  • Not affected by humidity
  • Suitable for micro-filtration due to fine fibers
  • High-loading capacity
  • High chemical purity
  • made with demineralized water
  • High permeability to air
Applications:
  • Filtration of industrial water, fresh water algae, drinking water, Proteins, samples for HPLC, solvents
  • Sediments content in petroleum products
  • Ligand binding test
  • Air pollution monitoring
  • Membranes pre-filters
  • Radio-immuno tests
  • Suspended solids in water, smokes
  • Liquid scintillation counting
  • Cells collection
  • Grease content in dairy products
Specifications

Specifications

75 g/m2
Disc
0.7 μm
Air pollution control, separation of soot before gas analyzers, support weighing in the balance analyzes and dryers
100 Pack
500°C
0.45 mm
55 mm
Microfilter
Borosilicate Glass
310 sec.
263
Non-hygroscopic
0.017 in.
Filter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers