Learn More
Mettler Toledo™ Accessory for Titrator: Titration Vessels
The right beaker for the right sample
1325.00€
Specifications
For Use With (Application) | Titration |
---|---|
Material | Glass |
For Use With (Equipment) | Titrator |
Description
- 09-313-151D: 250mL glass beakers for attachment directly to titration stand (set of 10)
- 09-313-151Q: 80mL glass beakers (set of 20)
Specifications
Titration | |
Glass | |
Titrator |