Learn More
Metro™ MetroMax i™ Top-Track Mobile Unit Kits
High-density, easy access storage
Brand:  Metro™ MXTTM18C
Additional Details : Weight : 12.16000kg
 View more versions of this product
Product Code. 15957221
  • 655.00€ / Each
Estimated Shipment: 27-09-2024
to see stock.
Add to basket
Includes: Posts, rigid casters, caster channels, bumpers and roller assemblies
Description
Description
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
  • Mobile units move along anchored track allowing easy access to all shelves
Order Info
  • Order one per mobile unit
  • Shelves sold separately
Specifications
Specifications
Posts, Chrome plated rigid casters, caster channel, bumpers and roller assemblies
Mobile Unit Kit
45.7 cm
18 in.
Metro™ MetroMax i™ and Q™ Top-Track™ system