Learn More
Metro™ High-Density qwikTRAK™ Mobile Unit Kits
Kit provides components for shelving units that fit on tracks for added mobility
Brand: Metro™ BTMS
Additional Details : Weight : 9.07200kg
Description
- For use with Metro Super Erecta™, Super Erecta Pro™ Super Adjustable™ and MetroMax iQ™ shelving systems
- Kit includes four 74 in. (188cm) high mobile posts, four V-groove casters; one stainless steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware
Order Info
Order one kit per mobile system. Shelves are sold separately.
Specifications
Four 74 in. high mobile posts, four V-groove casters, one Stainless Steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware | |
Mobile Unit Kit | |
Metro™ Super Erecta™, Super Adjustable™ shelving system |
11.4 kg | |
25 lb. | |
Stainless Steel |