Learn More
STEMCELL Technologies MethoCult™ SF H4236
Serum-free methylcellulose-based medium without cytokines for human cells
Brand: STEMCELL Technologies 04236
Additional Details : Weight : 0.08000kg
Description
- MethoCult™ SF H4236 is a serum-free, incomplete methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples
- This formulation does not contain fetal bovine serum, erythropoietin (EPO) or other cytokines and is recommended for testing cytokines and other supplements where the presence of serum is not desired
Specifications
Hematopoietic Stem and Progenitor Cells | |
Cell Culture, Colony Assay, Functional Assay | |
80 mL |