Learn More
Nickel Electro™ Metalware Range™ Stainless Steel Square Pipet Sterilizing Boxes
Keep pipets sterile with a sterilizing box. Nickel Electro™ Metalware Range™ Stainless Steel Square Pipet Sterilizing Boxes can be steam sterilized at 120°C and withstand dry heat temperatures of up to 180°C.
Brand: Nickel Electro™ 6704/SS
Additional Details : Weight : 1.00000kg
Description
- Stainless steel square profile
- Includes lid
- Square shape prevents it from rolling off bench top
- Silicon in lid and base
Specifications
Autoclavable | |
Stainless Steel | |
1 Ea. | |
Sterilizing Box |
490 x 70 mm | |
Sterile | |
490 mm | |
Autoclave |