missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury(Ii) Nitrate Monohydrate, purum p.a., ≥98.5% (T), Honeywell Fluka™
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Melting Point | 79°C |
Color | White |
Physical Form | Crystalline |
Quantity | 250 g |
Linear Formula | Hg(NO3)2 · H2O |
UN Number | UN1625 |
Formula Weight | 342.62g/mol |
Percent Purity | ≥98.5% (T) |
Packaging | Poly Bottle |
Chemical Name or Material | Mercury(II) nitrate monohydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'