Learn More
Taylor Wharton™ Measuring Rod
Click to view available options
:
Length: 96mm
Description
Order Info
LengthMetric
Specifications
Specifications
| Type | Measuring Rod |
| For Use With (Equipment) | Dipstick to assess the level of nitrogen remaining in the cryopreservative or tank. |
| Length (Metric) Exterior | 92 cm |