Learn More
MBL™ Borosilicate Glass Stirring Rod
Product Code. 12441627
Change view
Click to view available options
Length (Metric):
300 mm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
12441627 300 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12441627 Supplier MBL International Supplier No. SRF380
Trusted Glassware Deals
Use promo code 28509
View all promotions
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
Minimize scratching with these glass stirring rods. MBL™ Borosilicate Glass Stirring Rods are ideal for a wide range of stirring applications.
  • Borosilicate glass rods for general stirring applications
  • With rounded ends for extra safety and to minimize scratching
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) General stirring applications
Includes Rounded ends
Length (English) 11.81 in.
Diameter (Metric) 6 mm
Length (Metric) 300 mm
Material Borosilicate Glass
Quantity 10/Pk.