Learn More
Thermo Scientific™ Universal Clamps for MaxQ 4000, 4450, 6000, and Solaris Shakers
Mix and match these shaker platform clamps, mats and racks, available in a variety of sizes, to suit individual needs in the laboratory.
Brand:  Thermo Scientific™ 75004125
Additional Details : Weight : 2.81000kg
 View more versions of this product
Product Code. 17821905
  • 560.00€ / Each
Estimated Shipment: 31-05-2024
Description
Mix and match Thermo Scientific™ MaxQ™ Shaker Universal Clamps, available in a variety of sizes, to suit individual needs in the laboratory. Compatible with MaxQ shaker platforms.
Mix and match Thermo Scientific™ Solaris™ shaker Universal clamps to suit individual needs in the laboratory.
Specifications
Specifications
Separatory Funnel Clamp
403 x 159 x 174 mm platform
Separatory Funnel with 250 mL to 2 L Clamp
Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers