Learn More
Thermo Scientific™ Matrix™ Reagent Reservoirs
Product Code. p-4525611
Change view
Click to view available options
Capacity (Metric):
25 mL
75 mL
100 mL
Material:
Polypropylene
Polystyrene
Type:
Reagent Reservoir
Reagent Reservoir with Divider
Reservoir Lid
Unit Size:
Pack of 10
Pack of 100
Pack of 50
Packaging:
10 Bags of 10
10 pcs /bag
50 Individually wrapped
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Material Type Unit Size Packaging
10795393 75 mL Polypropylene Reagent Reservoir Pack of 10 -
10495973 75 mL Polypropylene Reservoir Lid Pack of 10 10 pcs /bag
10141873 100 mL Polystyrene Reagent Reservoir Pack of 100 10 Bags of 10
10375023 100 mL Polystyrene Reagent Reservoir Pack of 50 50 Individually wrapped
10756354 25 mL Polystyrene Reagent Reservoir Pack of 100 10 Bags of 10
10080263 25 mL Polystyrene Reagent Reservoir Pack of 50 50 Individually wrapped
10225743 25 mL Polystyrene Reagent Reservoir with Divider Pack of 100 10 Bags of 10
10435963 25 mL Polystyrene Reagent Reservoir with Divider Pack of 50 50 Individually wrapped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10795393 Supplier Thermo Scientific™ Supplier No. 8075
Add to basket
Edge
Add to basket
Accommodate a variety of applications with these uniquely designed, innovative reagent reservoirs.
Accomodate a variety of applications with uniquely designed, innovative Thermo Scientific™ Matrix Reagent Reservoirs. These sterile reservoirs offer an extra-wide base to add rigidity and stability, helping to avoid spills. The 25mL divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizing recovery if using only a single channel and a small amount of reagent.
  • Trough within a trough maximizes amount of liquid accessible to pipet tips when using small amounts of reagent
  • Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
  • Graduations on inside wall enable quick measurement of remaining liquid
  • Sterile reservoirs are packaged in snap and tear bags, maintaining sterility while being easy to open
TRUSTED_SUSTAINABILITY
Specifications
Color White
Capacity (Metric) 75 mL
Material Polypropylene
Sterility Non-sterile
Type Reagent Reservoir
No. per Case 10