Learn More
Masterflex™ Norprene™ Food I/P™ High Performance Precision Pump Tubing
Use this tubing for high-temperature food applications. Masterflex™ Norprene™ Food I/P™ High Performance Precision Pump Tubing is ideal for the food, dairy and beverage industries.
Brand: Masterflex™ 06402-89
Additional Details : Weight : 2.52000kg
Description
- For applications involving pressure, suction lift, viscous fluids or long tubing life
- Thicker wall compared to Precision pump tubing
- Heat, ozone and UV light resistant
- Non-aging and non-oxidizing
- Superior acid and alkali resistance
- Complies with FDA, 3A, NSF, REACH, RoHS
- Opaque beige
Specifications
Pump Tubing | |
15.9 mm | |
1500 cm | |
-51°C | |
89 | |
5/8 in. | |
Opaque Beige | |
610 mm |
Autoclavable | |
Norprene | |
food, dairy and beverage industries | |
Acids | |
0.03 to 19L/min. | |
8.2 m | |
20 psi | |
135°C |