Learn More
Thermo Scientific™ Manual Crimpers
Achieve secure, repeatable crimps or remove seals with minimized risk of damage with Thermo Scientific manual crimpers.
Brand:  Thermo Scientific™ C4020-103
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12410268
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
  • Lightweight, precision crimpers attach seals to chromatography or serum vials.
  • Available for standard aluminum or flip-off seals.
  • Factory set to properly crimp standard-thickness seals.
  • Adjustable handle setting screw allows proper crimps with septa of varying thickness.
Specifications
Specifications
Manual Crimper
20 mm
Autosampler Vial Closures