Learn More
Thermo Scientific™ Manual Crimpers
Achieve secure, repeatable crimps or remove seals with minimized risk of damage with Thermo Scientific manual crimpers.
Brand: Thermo Scientific™ C4020-103
Additional Details : Weight : 0.01000kg
Description
- Lightweight, precision crimpers attach seals to chromatography or serum vials.
- Available for standard aluminum or flip-off seals.
- Factory set to properly crimp standard-thickness seals.
- Adjustable handle setting screw allows proper crimps with septa of varying thickness.
Specifications
Manual Crimper | |
20 mm | |
Autosampler Vial Closures |