missing translation for 'onlineSavingsMsg'
Learn More

Magnesium bromide hexahydrate, extra pure, Thermo Scientific Chemicals

Catalog Number p-4813524
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
13446-53-2
Br2H12MgO6
292.20
MFCD00149780
LGLXXNHIGIJYQQ-UHFFFAOYSA-L
acmc-1bp1b, magnesium bromide-water 1/2/6, magnesium 2+ ion hexahydrate dibromide, magnesium 2+ hexahydrate dibromide, grignard reagent
17749097
O.O.O.O.O.O.Br[Mg]Br
This item is not returnable. View return policy
13446-53-2
Br2H12MgO6
292.20
MFCD00149780
LGLXXNHIGIJYQQ-UHFFFAOYSA-L
acmc-1bp1b, magnesium bromide-water 1/2/6, magnesium 2+ ion hexahydrate dibromide, magnesium 2+ hexahydrate dibromide, grignard reagent
17749097
O.O.O.O.O.O.Br[Mg]Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 165.0°C
Quantity 250 g
Linear Formula MgBr2·6H2O
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
IUPAC Name magnesium;dibromide;hexahydrate
Formula Weight 292.21
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Magnesium bromide hexahydrate
compliance-icons
Product Identifier
  • Magnesium bromide hexahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'