Learn More
Novus Biologicals™ MAGEB10 Overexpression Lysate
Brand:  Novus Biologicals™ NBP2-04861
Additional Details : Weight : 0.00970kg
Product Code. 18096256
  • 685.73€ / 0.10mg
Estimated Shipment: 15-07-2024
Description
The MAGEB10 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
Western Blot
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Native
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_872312. The protein contains a C-terminal DDK tag.
RIPA buffer
Homo sapiens MAGEB10, mRNA.
Cancer