Learn More
Macherey-Nagel™ Lysis Buffer T1
Lysis buffer for isolation and clean-up of genomic DNA samples using compatible kits (e.g., NucleoSpin™ Tissue kits, NucleoMag™ DNA Food).
Brand: Macherey-Nagel™ 740940.100
Additional Details : Weight : 0.10000kg
Specifications
Colorless | |
Compatible kits (e.g. NucleoSpin Tissue kits, NucleoMag DNA Food) | |
7.5 to 8.5 | |
Odorless | |
Liquid | |
Lysis Buffer T1 |
Up to 2% | |
Chemicals mixture in water | |
100 mL | |
Room Temperature (18°C to 25°C) | |
Isolation and cleanup of DNA |