Learn More
MP Biomedicals™ BigPrep™ Lysing Matrix E, 50 mL Tubes
BigPrep™ Lysing Matrix E contains ceramic spheres, silica spheres and glass beads for grinding soil, feces, sludge and other environmental samples.
28.41€ - 578.76€
Specifications
Bead Type | 1.4 mm Ceramic Spheres, 0.1 mm Silica Spheres, 4 mm Glass Beads |
---|---|
Content And Storage | Room Temperature |
Sample Type | Soil, Environmental, Feces |
DNase | Dnase-free |
For Use With (Equipment) | FastPrep instruments or other bead beaters |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15702472
|
MP Biomedicals™
116954002 |
2 x 50 mL Tubes |
28.41€
Pack of 2 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
11431565
|
MP Biomedicals™
116954010 |
10 x 50 mL Tubes |
138.86€
Pack of 10 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
11441565
|
MP Biomedicals™
116954050 |
50 x 50 mL Tubes |
578.76€
Pack of 50 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Lysing Matrix E tubes contain 1.4 mm ceramic spheres, 0.1 mm silica spheres, and 4 mm glass beads. They are used with FastPrep instruments or any bead beater to lyse and grind environmental samples for nucleic acid extraction. Lysing Matrix E is also effective for lysing mixed samples, such as microbe infected tissue for RNA extraction from tumors and other difficult tissues.
- Tube Size: 50 mL
- Sample Types: Soil, feces, sediment, wastewater, sludge, compost, rhizosphere
- Matrix Composition: 1.4 mm ceramic spheres, 0.1 mm silica spheres, and twenty-five 4 mm glass beads
Specifications
1.4 mm Ceramic Spheres, 0.1 mm Silica Spheres, 4 mm Glass Beads | |
Soil, Environmental, Feces | |
FastPrep instruments or other bead beaters | |
Lysing Matrix E | |
Up to 7 g per prep |
Room Temperature | |
Dnase-free | |
Nucleic Acid Isolation and Purification, Homogenization | |
RNase-free |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.