Learn More
MP Biomedicals™ Lysing Matrix B, 96-well Rack
Lysing Matrix B, 96-tube Rack is pre-filled with 0.1 mm silica spheres and used to lyse bacteria and spores.
217.30€ - 1894.22€
Specifications
Bead Type | Silica Spheres |
---|---|
Content And Storage | Store at Room Temperature. |
Sample Type | Bacteria, Fungi, Soil, Animal Tissues |
DNase | Dnase-free |
For Use With (Equipment) | FastPrep-96 or other bead beaters |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15113063
|
MP Biomedicals™
116981010 |
10 Pk. |
1894.22€
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
15212870
|
MP Biomedicals™
116981001 |
1 Ea. |
217.30€
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
Description
Lysing Matrix B, 96-tube rack comes with 1.2 mL cluster-strip tube and cap assemblies pre-filled with 0.1 mm silica spheres. Use with FastPrep-96 instrument or similar bead beater.
- Tube size: 96-tube rack
- Sample types: Gram positive and negative bacteria, fungal tissues, spores
- Matrix composition: 0.1 mm silica spheres
Specifications
Silica Spheres | |
Bacteria, Fungi, Soil, Animal Tissues | |
FastPrep-96 or other bead beaters | |
1.2 mL Cluster-strip Tube and Cap Assemblies | |
RNase-free |
Store at Room Temperature. | |
Dnase-free | |
Nucleic Acid Isolation and Purification, Homogenization | |
Lysing Matrix B | |
Less than 80 mg per prep |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.