Learn More
Poulten & Graf GmbH Sanitex glass and metal syringe
Constructed from metal and glass. Poulten Graf™ Sanitex glass and metal syringes are perfect for measuring and dispensing liquids.
205.00€ - 236.00€
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Glass, Metal |
Product Type | Syringe |
Max. Temperature (Metric) | 134°C |
Quantity | 1/Ea. |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10321702
|
Poulten & Graf GmbH
7 20237 |
10 mL |
205.00€
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
10427461
|
Poulten & Graf GmbH
7 20241 |
20 mL |
236.00€
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
Description
Made of metal and glass
Amber graduations
Specifications
Autoclavable | |
Syringe | |
1/Ea. |
Glass, Metal | |
134°C |