Learn More
Corning™ LSE™ Nutating Mixer, EU Plug
Product Code. 17700799 Shop All Corning Products
Change view
Click to view available options
Plug Type:
EU Plug
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type Unit Size
17700799 EU Plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17700799 Supplier Corning™ Supplier No. 6721
  • ONLINE EXCLUSIVE PRICE
    362.00€
    337.56€ / Pack of 1
    Save 24.44€
    6% Off
Estimated Shipment: 06-01-2027
Add to basket
Add to basket
A small, personal-sized 3D mixer that is ideal for ambient and cold room experiments. Ideal for applications such as immunohistochemistry, mixing and resuspending small containers in blood or centrifuge tubes securely held to the mixer with a dimpled mat.
  • Small footprint
  • Cold room compatible
  • Dimpled Mat
  • 3D Motion
  • Ideal for mixing and resuspending samples
TRUSTED_SUSTAINABILITY
Specifications
Load Bearing Capacity (Metric) 0.8 kg
Tilt Angle 20°
Voltage 120 V
Temperature Range (Metric) 4°C to 40°C
Amperage 0.03 A
Load Bearing Capacity (English) 1.75 lb.
Certifications/Compliance cETLus
Plug Type EU Plug
Frequency 50/60 Hz
Depth (English) 5.75 in.
Depth (Metric) 14.6 cm
Dimensions (LxW) Platform 24 x 17 cm (9.5 x 6.75 in.)
Humidity Range ≤80%, Non-condensing
Includes Dimpled Mat
Warranty and Service Offering 2 Years