Learn More
Lovibond™ Conformance Filter Sets
Glass Standards
1135.00€
Specifications
Certifications/Compliance ISO 9001: 2000 quality management system
For Use With (Application) Allow quick and simple conformance check
Material Glass
View More Specs
Products 1
Product Code Brand For Use With (Equipment) Packaging Type Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Packaging Type Price Quantity & Availability  
12831993
View Documents Promotion Details
Lovibond™
139510
PFX-195/2, PFX1-195/2, PFX-880/P, PFXi-880/P, PFX-950/P, PFXi-950/P, PFX-995/P, PFXi-995/P colorimeters 3 Filters of 0.5, 3.5, 5 values ASTM Color Filter Set This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
Description
Description
Allow quick and simple conformance checks on Lovibond instruments. Each filter set consists of colored glasses representing a spread of colors from the scale of interest, which are mounted in suitable holders. They are supplied in a presentation box with a Certificate of Conformity that states the actual values for each filter and confirms that they have been manufactured and inspected under the control of an ISO 9001: 2000 Quality Management System. In addition,Tintometer's ISO 17025 accredited calibration laboratory can supply glass filters for Lovibond instruments with color measurements that are directly traceable to national standards.
Note: Conformance filters are assigned a nominal value and Lovibond Tintometer always endeavors to match this value as closely as possible. On occasion, because of the unique manner of the glass making process, it may not be possible to match the requested values exactly. The values achieved and the performance tolerances expected will, of course, always be reported.
Specifications
Specifications
ISO 9001: 2000 quality management system
Allow quick and simple conformance check
Glass