Learn More
Lovibond™ CHECKIT™ Comparator Test Kits: Phosphate
Accurate test kits for routine water analysis
Brand: Lovibond™ 147250
Additional Details : Weight : 0.25000kg
Includes:
- Aqua Comparator Housing
- Aqua Comparator Disc
- Phosphate HR Tablets (100 pack)
- 2 x 13.5 mm Sample Cells
- Tablet Crusher
- Instructions
- Carrying Case
Description
Accurate test kits for routine water analysis
- Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value
Specifications
±5% | |
Aqua comparator™ housing, aqua comparator™ disc, phosphate HR tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case | |
0 to 80mg/L |
Routine water analysis | |
100 | |
Phosphate Test Kit |
Safety and Handling
- phosphate HR
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
Hazard Statement
- H318-Causes serious eye damage.
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : sodium hydrogen sulphate, ammonium chloride, boric acid, ammonium meta-vanadate