Learn More
Lovibond™ Aqua Comparitor Color Discs: Chlorine
For testing water with the Aqua Comparator
26.10€
Specifications
For Use With (Application) | Testing water with the Aqua comparator™ |
---|---|
For Use With (Equipment) | Aqua comparator™ |
Product Code | Brand | Type | Range | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Range | Price | Quantity & Availability | ||||
11765438
|
Lovibond™
146000 |
Chlorine Color Disc | 0.02 to 0.3mg/L |
26.10€
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
11775438
|
Lovibond™
146010 |
Chlorine Color Disc | 0 to 1mg/L |
26.10€
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
11705448
|
Lovibond™
146040 |
Chlorine Color Disc | 0.1 to 2mg/L |
26.10€
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
11795438
|
Lovibond™
146030 |
Chlorine Color Disc | 10 to 300mg/L |
26.10€
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
11785438
|
Lovibond™
146020 |
Chlorine Color Disc | 0 to 4mg/L |
26.10€
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
11705528
|
Lovibond™
146330 |
Chlorine Dioxide Color Disc | 0.01 to 0.2mg/L |
26.10€
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for chlorine using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Testing water with the Aqua comparator™ | |
Aqua comparator™ |