Learn More
Tork Long-Lasting Multipurpose Cleaning Cloth W8
GREENER_CHOICE
Product Code. 30243838 Shop All Tork Products
Change view
Click to view available options
Color:
Blue
Green
Red
Yellow
Unit Size:
Pack of 8
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30243838 Blue Pack of 8
30243790 Yellow Pack of 8
30243877 Green Pack of 8
30243860 Red Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30243838 Supplier Tork Supplier No. 194450
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
In Stock
Add to basket
Edge
Add to basket
A durable and absorbent cleaning cloth that can be used repeatedly without tearing. It is available in four colours to prevent cross-contamination.
  • Wipe away stubborn spills with our strong and absorbent cleaning cloths.
  • Prevent cross-contamination by using different-coloured cloths for different jobs.
  • No need to change your cleaning routine - these cloths can be used with most solvents, disinfectants and detergents.
  • Avoid waste with strong and durable cloths that won't tear easily and can be reused.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleaning Cloth
Length (English) 15.16 in.
Length (Metric) 38.5 cm
Width (English) 11.81 in.
Width (Metric) 30 cm
Certifications/Compliance FSC MIX Credit, ISO 9001, ISO 14001
Color Blue
Packaging Quantity 320/Cs.
Ply 1-ply
Certifications/Compliance (Europe) FSC MIX Credit, ISO 9001, ISO 14001