missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Rhodamine Antibody Labeling Kit
Label antibodies, proteins, peptides and other biomolecules with rhodamine, using simple procedure. Set up conjugations in seconds, and use them within minutes.
527.88€ - 1035.84€
Specifications
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
---|---|
Product Type | Antibody Labeling Kit |
Alert:
For Research Use Only
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18048605
|
Novus Biologicals™
311-0015 |
1 mg |
1035.84€
1mg |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||||
18036005
|
Novus Biologicals™
311-0010 |
300 μg |
718.23€
1 set |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||||
18027913
|
Novus Biologicals™
311-0030 |
30 μg |
527.88€
1 set |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||||
Description
- No desalting or dialysis steps
- Lightning-link Rapid technology can be used to label small quantities of antibody with 100% recovery
- Rhodamine used in kit has absorption peaks at 520 and 544nm
- Maximum emission is obtained when the rhodamine is excited at 544nm
Specifications
Store at -20°C. Avoid freeze-thaw cycles. | |
Antibody Labeling Kit |