Learn More
Abeba™ Light 1151 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Brand:  Abeba™ 1151/43
Additional Details : Weight : 0.95900kg
 View more versions of this product
Product Code. 15687117
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • CE EN ISO 20347:2012 O2 FO SRA
  • Smooth leather white
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3559)
  • Sanitized treated
  • Antistatic
  • Temperature regulating special lining Comfortemp™
  • Instep area with lacing
Specifications
Specifications
CE EN ISO 20347:2012 O2 FO SRA
Leather
Slip-resistant
1151
White
43
Polyurethane