Learn More
Abeba™ Light 1151 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Brand: Abeba™ 1151/46
Additional Details : Weight : 0.95900kg
Description
- CE EN ISO 20347:2012 O2 FO SRA
- Smooth leather white
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3559)
- Sanitized treated
- Antistatic
- Temperature regulating special lining Comfortemp™
- Instep area with lacing
Specifications
CE EN ISO 20347:2012 O2 FO SRA | |
Leather | |
Slip-resistant | |
1151 |
White | |
46 | |
Polyurethane |