Learn More
Abeba™ Light 1120 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Brand:  Abeba™ 1120/42
Additional Details : Weight : 1.04000kg
 View more versions of this product
Product Code. 15656947
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • CE EN ISO 20347:2012 O1 FO SRA
  • Smooth leather, perforated
  • White
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3559)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Double velcro fastening
  • Reflective material
Specifications
Specifications
CE EN ISO 20347:2012 O1 FO SRA
Leather
Slip-resistant
1120
White
42
Polyurethane