Learn More
Abeba™ Light 1058 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Brand: Abeba™ 1058/36
Additional Details : Weight : 1.11900kg
Description
- CE EN ISO 20345:2011 S1 SRA
- Leather with Air-Mesh textile inlays
- White/gray
- Steel toe cap
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3557)
- Sanitized treated
- Antistatic
- Breathable lining with Silver ions
- Instep area with lacing
Specifications
CE EN ISO 20345:2011 S1 SRA | |
Leather | |
Slip-resistant | |
1058 |
White, Gray | |
36 | |
Polyurethane | |
Steel |