Learn More
Abeba™ Light 1055 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Brand:  Abeba™ 1055/37
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15676807
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • CE EN ISO 20345:2011 S1 SRA
  • Leather with Air-Mesh textile inlays
  • Black
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3557)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver Point
  • Instep area with lacing
  • Reflective material
  • Suitable for heel and foot lift
Specifications
Specifications
CE EN ISO 20345:2011 S1 SRA
Leather
Slip-resistant
1055
Black
37
Polyurethane
Steel