Learn More
Abeba™ Light 1051 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Brand: Abeba™ 1051/43
Additional Details : Weight : 1.11900kg
Description
- CE EN ISO 20345:2011 S2 SRA
- Smooth leather white
- Steel toe cap
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3557)
- Sanitized treated
- Antistatic
- Temperature regulating special lining Comfortemp™
- Instep area with lacing
Specifications
CE EN ISO 20345:2011 S2 SRA | |
Leather | |
Slip-resistant | |
1051 |
White | |
43 | |
Polyurethane | |
Steel |