Learn More
Abeba™ Light 1047 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Brand:  Abeba™ 1047/47
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15696787
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • CE EN ISO 20345:2011 S2 SRA
  • Smooth leather white
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3557)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver Point
  • Adjustable instep area
  • Double velcro fastening
  • Suitable for heel and foot lift
  • Overcap leather/ carbon
Specifications
Specifications
CE EN ISO 20345:2011 S2 SRA
Leather
Slip-resistant
1047
White
47
Polyurethane
Steel