Learn More
Abeba™ Light 1030 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Brand:  Abeba™ 1030/41
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15636717
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
  • CE EN ISO 20345:2011 SB A, E, FO SRA
  • Smooth leather white
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3556)
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Breathable lining with Silver ions
  • Instep area with elastic band
Specifications
Specifications
CE EN ISO 20345:2011 SB A, E, FO SRA
Leather
Slip-resistant
1030
White
41
Polyurethane
Steel