Learn More
Abeba™ Light 1028 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Brand:  Abeba™ 1028/43
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15676687
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
  • CE EN ISO 20345:2011 S2 SRA
  • Microfiber
  • White
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3557)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with elastic band
Specifications
Specifications
CE EN ISO 20345:2011 S2 SRA
Microfiber
Slip-resistant
1028
White
43
Polyurethane
Steel