Learn More
Ley Rubber Rubber Flask Adapter
Product Code. 12378845 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Description:
50 x 24 x 36mm
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
12378845 50 x 24 x 36mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12378845
  • ONLINE EXCLUSIVE PRICE
    16.77€
    10.91€ / Pack of 5
    Save 5.86€
    34% Off
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
Suitable for use with: Gooch funnels, sintered glass funnels, slit sleeve funnels and porcelain funnels.
  • Pliable, molded rubber sleeve to fit the conical part of Gooch crucible adapters and small Buchner funnels and make a vacuum-tight joint with the rim of a filter flask
Specifications
Description 50 x 24 x 36mm
Material Rubber
For Use With (Equipment) Gooch funnels, sintered glass funnels, slit sleeve funnels and porcelain funnels
No. per Pack 5
Product Type Flask Adapter