Learn More
Semadeni™ LDPE Containers
3.60€ - 12.15€
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11832642
|
Semadeni™
870 |
0.5 L |
3.60€
Each Qty Price Break Available |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
11842642
|
Semadeni™
871 |
1 L |
5.94€
Each Qty Price Break Available |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
11852642
|
Semadeni™
874 |
2 L |
12.08€
Each Qty Price Break Available |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
11862642
|
Semadeni™
875 |
3.5 L |
12.15€
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Specifications
Container | |
1 Each |