missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ LATEX 400nm Black Mini Antibody Conjugation Kit

The LATEX™ Conjugation Kit allows antibodies or proteins to be conjugated to high quality 400 nm black latex beads quickly and easily.

533.41€ - 883.12€

Specifications

Content And Storage -20°C
Target LATEX™ 400nm Black Mini
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18054624
View Documents Promotion Details
Novus Biologicals™
1004-0040
16 μg
533.41€
Pack of 4
View alternative products
Save Up to 
»
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
 
18005844
View Documents Promotion Details
Novus Biologicals™
1004-0100
40 μg
883.12€
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
 
Description

Description

The latex beads are designed for ease of use and have much improved handling compared with traditional latex. This has been achieved by specially treating the particles. The conjugation is covalent, and requires less antibody than traditional passive and covalent latex conjugates.

Kit Contents:

  • 1 vial of 10x Quencher buffer
  • 1 vial of Resuspension buffer
  • 1 vial of Reaction buffer B
  • 1 vial of Reaction buffer A
  • 4 or 10 Mini vials of 400nm Black LATEX™
Specifications

Specifications

-20°C
LATEX™ 400nm Black Mini
Videos
SDS
Documents

Documents

Product Certifications