Learn More
Kimberly-Clark™ Kleenex™ Standard Roll Toilet Tissue
Artikelnummer. 10010962
Ansicht ändern
Click to view available options
Ply:
2-ply
3-ply
Packaging Quantity:
6 x 6 Rolls x 350 Sheets
6 x 6 Rolls x 600 Sheets
8 x 8 Rolls x 350 Sheets
Packungsgröße:
Pack of 36
Pack of 64
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Ply Packaging Quantity Packungsgröße
10010962 2-ply 6 x 6 Rolls x 600 Sheets Pack of 36
10797724 2-ply 8 x 8 Rolls x 350 Sheets Pack of 64
12681466 3-ply 6 x 6 Rolls x 350 Sheets Pack of 36
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 10010962 Lieferant Kimberly-Clark™ Lieferanten-Nr. 8441
  • ONLINE EXCLUSIVE PRICE
    116.00€
    64.81€ / Pack of 36
    Save 51.19€
    44% Off
In Stock
Zum Warenkorb hinzufügen
Edge
Zum Warenkorb hinzufügen
Provides the comfort of home. Kleenex™ Standard Roll Toilet Tissue dissolves in cold water and will not clog drains.
  • Super-white, perforated and hygienic
  • Soft and absorbent
  • Dissolves in cold water so that the sewer pipes will not clog up
  • Perforation allows economical consumption, suitable for cost savings and reducing the amount of waste.
  • ISO 14001, ISO 9001 and FSC certified
  • Guaranteed responsible manufacturing processes and high quality
  • Ideal for washrooms in office buildings and hotels that value a noble image
TRUSTED_SUSTAINABILITY
Spezifikation
Product Type Toilet Tissue Roll
Length (English) 4.7 in.
Length (Metric) 12 cm
Width (English) 3.7 in.
Width (Metric) 9.3 cm
Material Paper
Certifications/Compliance ISO 14001, ISO 9001, EU Ecolabel and FSC certified
Color White
Dimensions (L x W) 12 x 9.3 cm (4.7 x 3.7 in.)
Format Standard perforated roll
For Use With (Application) Ideal for washrooms in office buildings and hotels, which put on a noble image value
Packaging Quantity 6 x 6 Rolls x 600 Sheets
Ply 2-ply
Mehr anzeigen Weniger anzeigen