Learn More
Kimberly-Clark™ Kimtech™ Pure W4 Wipers, 5 Packs x 100 White Cloths
Product Code. 12685606 Shop All Kimberly-Clark Professional Products
Change view
Click to view available options
Dimensions (L x W):
30.4 x 30.4 cm
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
12685606 30.4 x 30.4 cm Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12685606 Supplier Kimberly-Clark™ Supplier No. 7605
  • ONLINE EXCLUSIVE PRICE
    375.00€
    194.86€ / Pack of 500
    Save 180.14€
    48% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Suitable for ISO Class 4 or higher cleanroom environments. Perfect for dry wiping critical equipment and surfaces in pharmaceutical, biotechnology, medical device and food processing industries. 30.4cm (W). Double bagged with case liner.
  • 5 packs x 100 white cloths (500 total)
  • Low linting, 100% Polypropylene white base sheets
  • Perfect for pharmaceutical, high-tech biotechnology, medical device and food processing industries
  • Suitable for ISO Class 4, or higher, cleanroom environments
  • Acid, base and solvent resistant
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Wiper
Length (English) 12.59 in.
Length (Metric) 32.7 cm
Width (English) 12.59 in
Width (Metric) 32.7 cm
Material Paper
Color White
Dimensions (L x W) 30.4 x 30.4 cm
Packaging Quantity 100 Sheets x 5 Bags (500 total)