Learn More
DWK Life Sciences Kimble™ Low-Form Color-Comparison Tubes
Shadowless bottom construction allows for viewing with undistorted light
336.00€
Specifications
Material Borosilicate Glass
View More Specs
Products 1
Product Code Brand Volume (Metric) Graduations Price Quantity & Availability  
Product Code Brand Volume (Metric) Graduations Price Quantity & Availability  
11848512
View Documents Promotion Details
DWK Life Sciences
45310-50100
100 mL 50 and 100 mL
336.00€
Pack of 6
View alternative products
Save Up to 
»
Estimated Shipment: 16-10-2024
Description
Shadowless bottom construction allows for viewing with undistorted light
  • Rings and legend are durable white ceramic enamel
  • Rings coincide with specific volumes of liquid
  • Matched or unmatched sets of 50 and 100mL
  • Glass, nonsterile, autoclavable
Order Info
Graduated ring scale of any of the six tubes will not vary by more than 3mm from that of any other tube within the set.
Specifications
Specifications
Borosilicate Glass