Learn More
DWK Life Sciences Kimble™ Kontes™ Microcaps™ TLC Spotting Capillaries
Product Code. p-3208057
Change view
Click to view available options
Volume (Metric):
0.5, 1, 2, 5, 10 μL
100 μL
20 μL
5 μL
Unit Size:
Pack of 100
Pack of 500
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
13005813 5 μL Pack of 100
13025813 20 μL Pack of 100
13065813 100 μL Pack of 100
13075813 0.5, 1, 2, 5, 10 μL Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 13005813 Supplier DWK Life Sciences Supplier No. 7645000005
For spotting preadsorbent TLC plates
Precision-bore glass capillary tubes, cut to predetermined lengths, so that each capillary tube will hold a known volume of fluid when filled.
  • Capillary action draws fluid into tube
  • Dispense fluid by squeezing bulb
  • No unusual skills or training required
  • Immediate, professional results
  • Supplied in packages containing one bulb assembly and one dispenser vial with 100 micropipets
  • Volumetric tolerance: ±1%
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP type I, ASTM E438
Diameter (English) Inner 0.0174 in.
Diameter (English) Outer 0.0355 in.
Diameter (Metric) Inner 0.4419 mm
Diameter (Metric) Outer 0.9017 mm
Material Borosilicate Glass
Includes 1 Bulb assembly and 1 dispenser vial
Length (English) 1.25 in.
Packaging Type Case of 100 glass tubes
Volume (Metric) 5 μL
Length (Metric) 32 mm
Type Microcaps™ TLC Spotting Capillary