Learn More
DWK Life Sciences Kimble™ Kontes™ Extraction Manifold Replacement Components
For extraction manifold.
Brand:  DWK Life Sciences 971101-0347
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10432635
  • 185.00€ / Each
Estimated Shipment: 23-05-2024
Description
  • For use with Kontes SPE Manifold Sets (971000-1047, -3047, or -6047)
  • Kontes SPE Glassware and Manifolds are specially designed as support systems for 47mm and 90mm dia. solid phase extraction disks to give you the highest possible recoveries of extracted analytes from your drinking water, surface water and waste water samples
Specifications
Specifications
300 mL
Glass Funnel (300 mL)
Glass
4.7 cm
Solid Phase Extraction Manifold
47 mm (300 mL) Funnel
1.85 in.
47 mm