Learn More
DWK Life Sciences Kimble™ KIMAX™ Colorware™ Flasks
Colorful enamel graduations enhance readability and indicate approximate volumes
Brand: DWK Life Sciences 26500Y50
Additional Details : Weight : 0.80000kg
Description
Colorful enamel graduations enhance readability and indicate approximate volumes
- Narrow mouth
- Durable, matte-finish marking spot provides good writing surface
- Without stoppers
Compliance
Designed from ASTM™ E 1404, Type I, Class 1 requirements
Specifications
Rubber | |
1.96 in. | |
Borosilicate Glass | |
10 mL | |
7.8 cm | |
Narrow | |
1 |
Sunny Yellow Ceramic Enamel scale graduations | |
50 mL | |
20 to 50 mL | |
3.07 in. | |
Matte Finish Marking | |
50 mm |