Learn More
DWK Life Sciences Kimble™ 51 Expansion Borosilicate Glass Melting Point Capillaries
51 expansion borosilicate glass
Brand:  DWK Life Sciences 34500-99
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11853063
  • 336.00€ / Pack of 20
Estimated Shipment: 21-05-2024
Description
  • O.D.: 1.5 to 1.8mm
  • 100 capillaries per vial
Specifications
Specifications
0.25 mm
Borosilicate Glass
Both ends open
3.93 in.
Melting Point Capillary
USP type I, ASTM E438
1.5 to 1.8 mm
100 mm