Learn More
Kartell™ Kartell™ Polypropylene Micropipet Tips
Made from Polypropylene. Kartell™ Polypropylene Micropipet Tips are designed with high clarity and excellent water-repellent properties to keep the sample retention within the tip to a minimum.
Brand:  Kartell™ 0095700
Additional Details : Weight : 0.25000kg
Product Code. 10478832
  • 225.00€ / Pack of 10000
Estimated Shipment: 08-07-2024
Description
  • Compatibility: Kartell™, Gilson™,Eppendorf™,Biohit™,Hamilton™
  • Cadmium-free polypropylene
  • Water-repellent properties
  • CE and IVD certifications
Specifications
Specifications
200 to 1000 mL
CE and IVD
Gilson™, Eppendorf Pipetter, Sartorius™ Biohit™, Hamilton™
Blue
Bulk