Learn More
Kartell™ Kartell™ Polypropylene Micropipet Tips
Made from Polypropylene. Kartell™ Polypropylene Micropipet Tips are designed with high clarity and excellent water-repellent properties to keep the sample retention within the tip to a minimum.
Brand: Kartell™ 0095700
Additional Details : Weight : 0.25000kg
Description
- Compatibility: Kartell™, Gilson™,Eppendorf™,Biohit™,Hamilton™
- Cadmium-free polypropylene
- Water-repellent properties
- CE and IVD certifications
Specifications
200 to 1000 mL | |
CE and IVD | |
Gilson™, Eppendorf Pipetter, Sartorius™ Biohit™, Hamilton™ |
Blue | |
Bulk | |
Polypropylene |