Learn More
Hanna Instruments™ K-Type Penetrating Thermocouple Probe for KEY™ Pocket Thermometer
Provides a fast response time and improved accuracy over the entire range. Hanna Instruments™ K-Type Penetrating Thermocouple Probe for KEY™ Pocket Thermometer is a 130mm (5.1 in.) AISI 316 stainless steel probe with a sharp tip for penetration of semisolids.
Brand: Hanna Instruments™ HI98517-13
Additional Details : Weight : 0.80000kg
Description
The HI98517 KEY™ is a pocket thermometer with an interchangeable probe for quick spot measurements. With a fast response time, KEY™ is ideal for quality control and industrial temperature monitoring.
- AISI 316 stainless steel, sharp tip probe for penetrating semi-solid samples
- HI98517-13 Penetrating Probe is supplied with the thermometer
- Can be replaced without buying a new thermometer
- Additional probes can be exchanged for more accurate measurements in liquids and on surfaces
Specifications
Temperature monitoring in semi-solids | |
5.1 in. | |
Stainless Steel | |
1°C/°F | |
±2°C/4°F |
Max. 95% | |
130 mm | |
∼20 sec. in water | |
HI98517 KEY™ Pocket Thermometer |