Learn More
Emma Safety Footwear Jack S1P Denim Shoe
The unique heel design of the Jack Shoe is highly supportive and ideal for climbing pallets, ladders and in and out of truck cabs - even when wet.
Marke: Emma Safety Footwear MM91956941
Weitere Details : Weight : 1.08400kg
Beschreibung
- S1P-Safetyshoe with sole plate for dry work
- Metal Free
- Its unique heel design is highly supportive and ideal for climbing pallets, ladders and in and out of truck cabs - even when wet.
- The protective insole protects from sharp objects like nails and shards of metal.
- This sneaker with fiberglass toe cap, is finished with robust denim. DenimX laces with a story. Made from recycled jeans for our circular safety shoes.
- Industries:
- Logistics & transport
- Electronics
- Manufacturing
Spezifikation
ISO 20345:2011 | |
Blue | |
Hydro-Tec Antibacterial | |
41 | |
Low-cut |
Laces | |
Logistics and Transport, Electronics, Manufacturing | |
Denim, Nubuck | |
Extra-wide Width | |
Safety Shoe |