Learn More
Emma Safety Footwear Jack (Ruffneck) Safety Shoes
Sneaker with composite toe cap includes a unique rough neck heel design which is highly supportive and ideal for climbing pallets, ladders and getting in and out of truck cabs even when wet. The virtually impenetrable insole protects from sharp objects like nails and shards of metal.
Brand:  Emma Safety Footwear 912549-45
Additional Details : Weight : 1.66000kg
 View more versions of this product
Product Code. 15969061
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • S1P-Safety shoe with sole plate for dry work
  • Denim/Nubuck construction
  • GripForce™ Easy Twist Outsole
  • 3M™ Reflective Striping
  • ESD certified
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Specifications
EN ISO 20345:2011
Blue
Hydro-Tec™ Sanitized Silver
45
Laces
Jack (Ruffneck)
Denim, Nubuck
Composite