Learn More
GPE Scientific™ J Young Thumb Screw Lock Clips for Spherical Joints
GPE Scientific™ J Young Thumb Screw Lock Clips for Spherical Joints lock in place to securely fasten J Young spherical joints.
Brand:  GPE Scientific™ TJC/29
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 15844904
  • 37.15€ / Each
Estimated Shipment: 17-07-2024
Description
  • Thumb screw design
  • Securely fasten J Young spherical joints
Specifications
Specifications
Thumb Screw Lock Clips for J Young Spherical Joints
28/12 and 28/15 size spherical joints